missing translation for 'onlineSavingsMsg'
Learn More
Abnova™ SSR1 Recombinant Protein Artikelnummer.: 16151012
Nur noch undefined auf Lager
Zum Warenkorb hinzufügen

Abnova™ SSR1 Recombinant Protein

Product Code. 16151012
25 μg, 25 Mikrogramm
missing translation for 'orderingAttributeHoverText'
Menge:
10 μg
25 μg
missing translation for 'unitSize'
10 Mikrogramm
25 Mikrogramm
This item is not returnable. View return policy

Artikelnummer. 16151012

Marke: Abnova™ H00006745P01.25ug

Nur noch undefined auf Lager
Zum Warenkorb hinzufügen
Nur noch undefined auf Lager
Zum Warenkorb hinzufügen
This item is not returnable. View return policy

ELISA, Western Blotting (Recombinant Protein), Antibody Production, Protein Array

Specifications

Zugriffsnummer AAH07710
Zur Verwendung mit (Anwendung) Antibody Production, Protein Array, ELISA, Western Blot
Zusammensetzung 50mM Tris HCl, 10mM reduced Glutathione, pH 8 in the Elution Buffer
Gen-ID (Entrez) 6745
Molekulargewicht 53.68
Name SSR1 (Human) Recombinant Protein (P01)
pH-Bereich 8
Vorbereitungsmethode In vitro wheat germ expression system
Reinigungsverfahren Glutathione Sepharose 4 Fast Flow
Qualitätskontrollen 12.5% SDS-PAGE Stained with Coomassie Blue.
Menge 25 μg
Quelle Wheat Germ (in vitro)
Immunogen QDLTEDEETVEDSIIEDEDDEAEVEEDEPTDLVEDKEEEDVSGEPEASPSADTTILFVKGEDFPANNIVKFLVGFTNKGTEDFIVESLDASFRYPQDYQFYIQNFTALPLNTVVPPQRQATFEYSFIPAEPMGGRPFGLVINLNYKDLNGNVFQDAVFNQTVTVIEREDGLDGETIFMYMFLAGLGLLVIVGLHQLLESRKRKRPIQKVEMGTSSQNDVDMSWIPQETLNQINKASPRRLPRKRAQKRSVGSDE
Lagerungsbedingungen Store at -80°C. Aliquot to avoid repeated freezing and thawing.
Gen-Alias DKFZp781N23103/FLJ14232/FLJ22100/FLJ23034/FLJ78242/FLJ93042/TRAPA
Gebräuchliche Bezeichnung SSR1
Gensymbol SSR1
Kreuzreaktivität Human
Spezies Wheat Germ (in vitro)
Rekombinant Recombinant
Proteinmarkierung GST
Form Solution
Show More Show Less
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title
Abnova™ SSR1 Recombinant Protein >

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.