missing translation for 'onlineSavingsMsg'
Learn More
Learn More
SS18L2 Rabbit anti-Human, Mouse, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Marke: Proteintech 11817-1-AP-20UL
Additional Details : Gewicht : 0.00010kg
Spezifikation
SS18L2 | |
Polyclonal | |
Unconjugated | |
SS18L2 | |
KIAA iso, SS18 like protein 2, SS18L2, SYT homolog 2 | |
Rabbit | |
Antigen Affinity Chromatography | |
RUO | |
26901, 51188 | |
Antibody | |
IgG |
Immunohistochemistry (Paraffin), Western Blot | |
0.26 mg/mL | |
PBS with 50% glycerol and 0.1% sodium azide; pH 7.3 | |
Q9D174, Q9UHA2 | |
SS18L2 | |
SS18L2 Fusion Protein Ag2384 | |
20 μL | |
Primary | |
Mouse, Human | |
Liquid |