missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ SRMS (SRM) Recombinant Human Protein
SRMS (Srm) is a tyrosine protein kinase (SRC sub-family).
2895.00 CHF - 18250.00 CHF
Spezifikation
Zugriffsnummer | NP_543013 |
---|---|
Zur Verwendung mit (Anwendung) | Enzyme and Protein Activity Assay, Kinase Assay, Protein Assays and Analysis, Protein Biology |
Zusammensetzung | 50mM HEPES pH 7.0, 150mM KCl, 0.5mM EDTA, 0.02% Triton X-100, 2mM DTT, 50% Glycerol |
Molekulargewicht | 82kDa |
Lagerungsbedingungen | Store at -80°C. At first use, aliquot and store at -80°C to avoid multiple freeze-thaws. |
Produktcode | Marke | Menge | Preis | Menge & Verfügbarkeit | |||||
---|---|---|---|---|---|---|---|---|---|
Produktcode | Marke | Menge | Preis | Menge & Verfügbarkeit | |||||
15488503
|
Invitrogen™
PR7031A |
100μg |
2895.00 CHF
100 Mikrogramm |
Verfügbar ab: 14-05-2024
Loggen Sie sich ein, um den Lagerbestand zu sehen |
|||||
10473843
|
Invitrogen™
PV4287 |
1mg |
18250.00 CHF
1 Milligramm |
Verfügbar ab: 14-05-2024
Loggen Sie sich ein, um den Lagerbestand zu sehen |
|||||
Beschreibung
SRMS (Srm) may be involved in the proliferation and differentiation of keratinocytes.
Spezifikation
NP_543013 | |
50mM HEPES pH 7.0, 150mM KCl, 0.5mM EDTA, 0.02% Triton X-100, 2mM DTT, 50% Glycerol | |
Store at -80°C. At first use, aliquot and store at -80°C to avoid multiple freeze-thaws. | |
Full Length | |
SRC Kinase Family | |
Recombinant | |
Baculovirus |
Enzyme and Protein Activity Assay, Kinase Assay, Protein Assays and Analysis, Protein Biology | |
82kDa | |
RUO - research use only | |
SRMS (Srm) | |
Human | |
GST Tag | |
Frozen |