missing translation for 'onlineSavingsMsg'
Learn More
Learn More
SR-BI Rabbit anti-Human, Mouse, Rat, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Marke: Proteintech 21277-1-AP-20UL
Additional Details : Gewicht : 0.00010kg
Beschreibung
Receptor for different ligands such as phospholipids, cholesterol ester, lipoproteins, phosphatidylserine and apoptotic cells. Probable receptor for HDL, located in particular region of the plasma membrane, called caveolae. Facilitates the flux of free and esterified cholesterol between the cell surface and extracellular donors and acceptors, such as HDL and to a lesser extent, apoB-containing lipoproteins and modified lipoproteins. Probably involved in the phagocytosis of apoptotic cells, via its phosphatidylserine binding activity. Receptor for hepatitis C virus glycoprotein E2. Binding between SCARB1 and E2 was found to be independent of the genotype of the viral isolate. Plays an important role in the uptake of HDL cholesteryl ester.Spezifikation
SR-BI | |
Polyclonal | |
Unconjugated | |
SCARB1 | |
CD36 and LIMPII analogous 1, CD36 antigen like 1, CD36L1, CLA 1, CLA1, SCARB1, SRB1, SR-B1, SR-BI | |
Rabbit | |
Antigen Affinity Chromatography | |
RUO | |
20778, 25073, 949 | |
-20°C | |
Liquid |
Western Blot, Immunocytochemistry, Immunofluorescence | |
0.43 mg/mL | |
PBS with 50% glycerol and 0.02% sodium azide; pH 7.3 | |
P97943, Q61009, Q8WTV0 | |
SCARB1 | |
SR-BI Fusion Protein Ag15759 | |
20 μL | |
Primary | |
Mouse, Rat, Human | |
Antibody | |
IgG |