missing translation for 'onlineSavingsMsg'
Learn More
Learn More
SPRY2 Rabbit anti-Human, Mouse, Rat, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Marke: Proteintech 11383-1-AP-20UL
Additional Details : Gewicht : 0.00010kg
Beschreibung
This gene encodes a protein belonging to the sprouty family. The encoded protein contains a carboxyl-terminal cysteine-rich domain essential for the inhibitory activity on receptor tyrosine kinase signaling proteins and is required for growth factor stimulated translocation of the protein to membrane ruffles. In primary dermal endothelial cells this gene is transiently upregulated in response to fibroblast growth factor two. This protein is indirectly involved in the non-cell autonomous inhibitory effect on fibroblast growth factor two signaling. The protein interacts with Cas-Br-M (murine) ectropic retroviral transforming sequence, and can function as a bimodal regulator of epidermal growth factor receptor/mitogen-activated protein kinase signaling. This protein may play a role in alveoli branching during lung development as shown by a similar mouse protein.Spezifikation
SPRY2 | |
Polyclonal | |
Unconjugated | |
SPRY2 | |
hSPRY2, Protein sprouty homolog 2, sprouty homolog 2 (Drosophila), Spry 2, SPRY2 | |
Rabbit | |
Antigen Affinity Chromatography | |
RUO | |
10253, 24064, 306141 | |
-20°C | |
Liquid |
Immunohistochemistry (Paraffin), Western Blot, Immunofluorescence, Immunocytochemistry | |
0.43 mg/mL | |
PBS with 50% glycerol and 0.1% sodium azide; pH 7.3 | |
O43597, Q9QXV8 | |
SPRY2 | |
SPRY2 Fusion Protein Ag1918 | |
20 μL | |
Primary | |
Rat, Human, Mouse | |
Antibody | |
IgG |