Learn More
Fisher Chemical 125 ML SPECTRAPURE - Optimisation Standard (XXII) 5 components -Cd, Cu, Mg, Pb, Rh 200µg/L of each
Marke: Fisher Chemical PUR051125
125 ML SPECTRAPURE - Optimisation Standard (XXII) 5 components -Cd, Cu, Mg, Pb, Rh 200µg/L of each
Additional Details : Gewicht : 0.12500kg