Learn More
SPATA32 Polyclonal Antibody, Invitrogen™
Rabbit Polyclonal Antibody
Marke: Thermo Scientific PA555104
Additional Details : Gewicht : 0.02000kg
Beschreibung
Immunogen sequence: SVHSNMGLPT PQTFRPWSLN SNCRSFTEEN HVSACHHSIS AQTSKHLFWA NKLIQASEHS LQRAINMQLN NGSAGQPIRS PL Highest antigen sequence identity to the following orthologs: Mouse - 37%, Rat - 39%.
SPATA32 Polyclonal antibody specifically detects SPATA32 in Human samples. It is validated for Immunohistochemistry.Spezifikation
SPATA32 | |
Polyclonal | |
Unconjugated | |
SPATA32 | |
acrosome expressed 2; AEP2; C17orf46; CTD-2020K17.4; SPATA32; spermatogenesis associated 32; spermatogenesis-associated protein 32; testis expressed 34; Testis-expressed protein 34; testis-expressed sequence 34 protein; TEX34; VAD1.2 | |
Rabbit | |
Antigen affinity chromatography | |
RUO | |
124783 | |
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
Liquid |
Immunohistochemistry | |
0.1 mg/mL | |
PBS with 40% glycerol and 0.02% sodium azide; pH 7.2 | |
Q96LK8 | |
SPATA32 | |
Recombinant protein corresponding to Human SPATA32 | |
100 μL | |
Primary | |
Human | |
Antibody | |
IgG |
Research Use Only