missing translation for 'onlineSavingsMsg'
Learn More
Learn More
SPANXC Rabbit anti-Human, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Marke: Proteintech 11957-1-AP-20UL
Additional Details : Gewicht : 0.00010kg
Beschreibung
Temporally regulated transcription and translation of several testis-specific genes is required to initiate the series of molecular and morphological changes in the male germ cell lineage necessary for the formation of mature spermatozoa. This gene is a member of the SPANX family, which is located in a gene cluster on chromosome X. The SPANX genes encode differentially expressed testis-specific proteins that localize to various subcellular compartments. This particular gene encodes a protein that localizes to the nucleus and is expressed in highly metastatic cell lines, making the protein a potential diagnostic and prognostic marker. The protein belongs to a family of cancer/testis antigens and represents a potential target for cancer immunotherapy.Spezifikation
SPANXC | |
Polyclonal | |
Unconjugated | |
SPANXC | |
C, Cancer/testis antigen 11.3, CT11.3, CTp11, SPANX C, SPANX family member C, SPANX family, member C, SPANXC | |
Rabbit | |
Antigen Affinity Chromatography | |
RUO | |
64663 | |
-20°C | |
Liquid |
Western Blot | |
0.5 mg/mL | |
PBS with 50% glycerol and 0.02% sodium azide; pH 7.3 | |
Q9NY87 | |
SPANXC | |
SPANXC Fusion Protein Ag2561 | |
20 μL | |
Primary | |
Human | |
Antibody | |
IgG |