missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Solute carrier family 22 member 18 Antibody, Novus Biologicals™
Rabbit Polyclonal Antibody
561.00 CHF
Spezifikation
| Antigen | Solute carrier family 22 member 18 |
|---|---|
| Anwendungen | Western Blot |
| Klassifikation | Polyclonal |
| Konjugat | Unconjugated |
| Wirtsspezies | Rabbit |
| Produktcode | Marke | Menge | Preis | Menge & Verfügbarkeit | |||||
|---|---|---|---|---|---|---|---|---|---|
| Produktcode | Marke | Menge | Preis | Menge & Verfügbarkeit | |||||
|
18264944
|
Novus Biologicals
NBP1-59747 |
100 μL |
561.00 CHF
100 Mikroliter |
Log in om dit product te kopen Registreer vandaag om een webaccount aan te maken | |||||
Beschreibung
Solute carrier family 22 member 18 Polyclonal specifically detects Solute carrier family 22 member 18 in Human samples. It is validated for Western Blot.Spezifikation
| Solute carrier family 22 member 18 | |
| Polyclonal | |
| Rabbit | |
| BWR1AORCTL2, BWSCR1Asolute carrier family 22 (organic cation transporter), member 1-like, Efflux transporter-like protein, HET, imprinted multi-membrane spanning polyspecific transporter-related protein 1, Imprinted multi-membrane-spanning polyspecific transporter-related protein 1, IMPT1DKFZp667A184, ITMp45-BWR1A, ORCTL-2, organic cation transporter-like 2, Organic cation transporter-like protein 2, p45 Beckwith-Wiedemann region 1A, SLC22A1Lcandidate A, solute carrier family 22 member 18, Solute carrier family 22 member 1-like, solute carrier family 22, member 18, TSSC5p45-Beckwith-Wiedemann region 1 A, Tumor-suppressing STF cDNA 5 protein, Tumor-suppressing subchromosomal transferable fragment candidate gene 5 protein | |
| SLC22A18 | |
| IgG |
| Western Blot | |
| Unconjugated | |
| RUO | |
| 5002 | |
| Synthetic peptides corresponding to SLC22A18(solute carrier family 22, member 18) The peptide sequence was selected from the N terminal of SLC22A18. Peptide sequence AASSPALPGVYLLFASRLPGALMHTLPAAQMVITDLSAPEERPAALGRLG. | |
| Primary |
Haben Sie Verbesserungsvorschläge?Übermitteln Sie eine inhaltliche Korrektur
Berichtigung von Produktinhalten
Bitte geben Sie uns Ihr Feedback zu den Produktinhalten, indem Sie das folgende Formular ausfüllen.
Name des Produkts