Learn More
Chem-Lab SOL COT STANDARD 1L 500PPM ORGANIC
Marke:  Chem-Lab CL01.2086.500.1000
SOL COT STANDARD 1L 500PPM ORGANIC
Artikelnummer. 11960409
  • ONLINE EXCLUSIVE PRICE
    183.00 CHF
    99.91 CHF / 1 Stück
    Sparen 83.09 CHF
    45% Rabatt