Learn More
Natriumsulfat-Decahydrat, zertifiziertes AR für Analysen, Fisher Chemical
Marke: Fisher Chemical S/6560/60
Additional Details : CAS : 7727-73-3 Gewicht : 1.00000kg
Menge | 1kg |
---|
Chemische Identifikatoren
7727-73-3 | |
322.186 | |
RSIJVJUOQBWMIM-UHFFFAOYSA-L | |
62649 | |
Dinatrium; Sulfat; Decahydrat |
H20Na2O14S | |
149189 | |
sodium sulfate decahydrate, glauber's salt, disodium sulfate decahydrate, sodium sulfate usp, sulfuric acid disodium salt, decahydrate, unii-0ypr65r21j, sodium sulfate na2so4 decahydrate, sodium sulphate decahydrate, sodium sulfate decahydrate na2so4.10h2o, disodium sulfate decahydrate na2so4.10h2o | |
CHEBI:32586 | |
O.O.O.O.O.O.O.O.O.O.[O-]S(=O)(=O)[O-].[Na+].[Na+] |
Spezifikation
7727-73-3 | |
149189 | |
RSIJVJUOQBWMIM-UHFFFAOYSA-L | |
Dinatrium; Sulfat; Decahydrat | |
62649 | |
322.2g/mol | |
Kunststoffpulverbehälter | |
884°C | |
1kg |
H20Na2O14S | |
sodium sulfate decahydrate, glauber's salt, disodium sulfate decahydrate, sodium sulfate usp, sulfuric acid disodium salt, decahydrate, unii-0ypr65r21j, sodium sulfate na2so4 decahydrate, sodium sulphate decahydrate, sodium sulfate decahydrate na2so4.10h2o, disodium sulfate decahydrate na2so4.10h2o | |
O.O.O.O.O.O.O.O.O.O.[O-]S(=O)(=O)[O-].[Na+].[Na+] | |
322.186 | |
CHEBI:32586 | |
Fest | |
Weiß | |
5.2 to 9.2 |