missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Natriumnitroferricyanid(Iii)-Dihydrat, Honeywell Fluka™
Click to view available options
Menge:
5 g
100 g
500 g
Beschreibung
- Leading the Industry For Over 65 Years
- Honeywell now delivers Fluka™ premium grade inorganic reagents worldwide – with consistency, purity, and accuracy assured
Spezifikation
Chemischer Name oder Material | Sodium Nitroferricyanide(Iii) Dihydrate |
Menge | 100 g |
Strukturformel | Na2[Fe(CN)5NO] · 2H2O |
UN-Nummer | 3288 |
IUPAC-Name | disodium pentacyano(oxidaniumylidyneamino)ironbis(ylium) dihydrate |
Formelmasse | 297.95g/mol |
Reinheit (%) | ≥99% |
Güte | ACS Reagent |
missing translation for 'provideContentCorrection'
missing translation for 'productTitle'