missing translation for 'onlineSavingsMsg'
Learn More

Natriumhydroxid-Lösung, 0.25 M, Honeywell Fluka™

Catalog Number 15653210
Click to view available options
Menge:
1 l
5 l
10 L
6 x 1 l
Dieser Artikel kann nicht zurückgegeben werden. Rückgaberichtlinie anzeigen
39.997
MFCD00003548
HEMHJVSKTPXQMS-UHFFFAOYSA-M
sodium hydroxide, caustic soda, sodium hydrate, white caustic, soda lye, aetznatron, ascarite, sodium hydroxide na oh, sodium hydroxide solution, soda, caustic
14798
CHEBI:32145
[OH-].[Na+]
Dieser Artikel kann nicht zurückgegeben werden. Rückgaberichtlinie anzeigen
39.997
MFCD00003548
HEMHJVSKTPXQMS-UHFFFAOYSA-M
sodium hydroxide, caustic soda, sodium hydrate, white caustic, soda lye, aetznatron, ascarite, sodium hydroxide na oh, sodium hydroxide solution, soda, caustic
14798
CHEBI:32145
[OH-].[Na+]

Volumetrisch, 0.25 M NaOH (0.25 N)

Honeywell´s high-quality titration products cover the complete titration workflow and include:

  • Solutions for volumetric titration (acids, bases, or salts)
  • Buffers available as concentrates or ready-to-use

 

  • Reagents for complexometry:
    • Aminopolycarboxylic acids (EDTA/NTA analogs)
    • Masking agents
  • Indicators for volumetric and complexometric titrations

 


 

TRUSTED_SUSTAINABILITY
Chemischer Name oder Material Sodium Hydroxide Solution
Strukturformel NaOH
Konzentration 0.25M
UN-Nummer NONH for all modes of transport
Menge 10 L
Dampfdruck 3 mmHg (37°C)
missing translation for 'provideContentCorrection'

missing translation for 'productTitle'