Learn More
Thermo Scientific™ Sodium cacodylate, 0.1M buffer soln., pH 7.0
Beschreibung
Sodium cacodylate,is used as an arsinate compound for proteomics research. It is used as a buffer in electron microscopy.
This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.
Spezifikation
Spezifikation
| Physikalische Form | Flüssigkeit |
| Menge | 100 mL |