missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Natriumbromid, Honeywell Fluka™
Click to view available options
Menge:
500 g
1 kg
2.5 kg
6 x 1 kg
Beschreibung
- Leading the Industry For Over 65 Years
- Honeywell now delivers Fluka™ premium grade inorganic reagents worldwide – with consistency, purity, and accuracy assured
Spezifikation
Chemischer Name oder Material | Sodium Bromide |
Namenshinweis | Meets analytical specification of Ph. Eur.BP,USP |
Menge | 6 x 1 kg |
Strukturformel | NaBr |
UN-Nummer | NONH for all modes of transport |
IUPAC-Name | sodium bromide |
Formelmasse | 102.89g/mol |
Reinheit (%) | 99 to 100.5% (calc. to the dried substance) |
Güte | puriss. |
Verpackung | Kunststoffflasche |
Mehr anzeigen |
missing translation for 'provideContentCorrection'
missing translation for 'productTitle'