Learn More
Duran™ Trichter aus Kalknatronglas
Used for decanting and filtration of substances
Marke: Duran™ 233513305
Additional Details : Gewicht : 0.20000kg
Beschreibung
- Suited to use with hot or aggressive media
- Good thermal shock and chemical resistance
- Typical applications include decanting and filtration of substances
Spezifikation
55 mm | |
Kalknatronglas | |
ISO 4798, USP-Standard | |
Zum Abgießen und Filtern von Substanzen |
8 mm | |
95 mm | |
70 bis 90 mm |