missing translation for 'onlineSavingsMsg'
Learn More
Learn More
SMNDC1 Rabbit anti-Human, Mouse, Rat, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Marke: Proteintech 12178-1-AP-150UL
Additional Details : Gewicht : 0.00010kg
Beschreibung
This protein is a paralog of SMN1 gene, which encodes the survival motor neuron protein, mutations in which are cause of autosomal recessive proximal spinal muscular atrophy. The protein encoded by this gene is a nuclear protein that has been identified as a constituent of the spliceosome complex. This protein is differentially expressed, with abundant levels in skeletal muscle, and may share similar cellular function as the SMN1 gene.Spezifikation
SMNDC1 | |
Polyclonal | |
Unconjugated | |
SMNDC1 | |
30 kDa splicing factor SMNrp, SMN related protein, SMNDC1, SMNR, SPF30 | |
Rabbit | |
Antigen Affinity Chromatography | |
RUO | |
10285, 287768, 76479 | |
-20°C | |
Liquid |
Immunohistochemistry (Paraffin), Western Blot | |
0.13 mg/mL | |
PBS with 50% glycerol and 0.1% sodium azide; pH 7.3 | |
O75940, Q4QQU6, Q8BGT7 | |
Smndc1 | |
SMNDC1 Fusion Protein Ag2822 | |
150 μL | |
Primary | |
Human, Mouse, Rat | |
Antibody | |
IgG |