missing translation for 'onlineSavingsMsg'
Learn More
Learn More
SMMHC Rabbit anti-Human, Mouse, Rat, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Marke: Proteintech 18569-1-AP-150UL
Additional Details : Gewicht : 0.00010kg
Beschreibung
The protein encoded by MYH11 is a smooth muscle myosin belonging to the myosin heavy chain family. The gene product is a subunit of a hexameric protein that consists of two heavy chain subunits and two pairs of non-identical light chain subunits. It functions as a major contractile protein, converting chemical energy into mechanical energy through the hydrolysis of ATP. The gene encoding a human ortholog of rat NUDE1 is transcribed from the reverse strand of MYH11, and its 3′ end overlaps with that of the latter. The pericentric inversion of chromosome 16 [inv(16)(p13q22)] produces a chimeric transcript that encodes a protein consisting of the first 165 residues from the N terminus of core-binding factor beta in a fusion with the C-terminal portion of the smooth muscle myosin heavy chain. This chromosomal rearrangement is associated with acute myeloid leukemia of the M4Eo subtype. Alternative splicing generates isoforms that are differentially expressed, with ratios changing during muscle cell maturation. Alternatively spliced transcript variants encoding different isoforms have been identified.Spezifikation
SMMHC | |
Polyclonal | |
Unconjugated | |
MYH11 | |
MYH11, Myosin 11, Myosin heavy chain 11, SMHC, SMMHC, SMM-HC, smooth muscle Myosin heavy chain | |
Rabbit | |
Antigen Affinity Chromatography | |
RUO | |
17880, 24582, 4629 | |
-20°C | |
Liquid |
Western Blot | |
0.16 mg/mL | |
PBS with 50% glycerol and 0.02% sodium azide; pH 7.3 | |
O08638, P35749, Q63862 | |
Myh11 | |
SMMHC Fusion Protein Ag13391 | |
150 μL | |
Primary | |
Human, Mouse, Rat | |
Antibody | |
IgG |