missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ SMC2 Polyclonal Antibody
GREENER_CHOICE

Artikelnummer. 17178323
Change view
Click to view available options
Menge:
100 μL
Packungsgröße:
100 Mikroliter
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Artikelnummer. Menge unitSize
17178323 100 μL 100 Mikroliter
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Dieser Artikel kann nicht zurückgegeben werden. Rückgaberichtlinie anzeigen
Artikelnummer. 17178323 Lieferant Invitrogen™ Lieferanten-Nr. PA5110399

om dit product te kopen Registreer vandaag om een webaccount aan te maken

Dieser Artikel kann nicht zurückgegeben werden. Rückgaberichtlinie anzeigen

Rabbit Polyclonal Antibody

Immunogen sequence: EELAGLKNTA EKYRQLKQQW EMKTEEADLL QTKLQQSSYH KQQEELDALK KTIEESEETL KNTKEIQRKA EEKYEVLENK MKNAEAERER ELKDAQKKLD CAKTKADASS KKMKEKQQEV EAITLELEEL KREHTSYKQQ LEAVNEAIKS YESQIEVMAA EVAKNKESVN KAQEEVTKQK EVITAQDTVI KAKYAEVAKH K.

CAP-E and CAP-C (Chromosome Associated Protein-E & C) are also SMC (Structural Maintenance of Chromosome) family members that form a heterodimeric complex required for mitotic chromosome condensation to achieve proper segregation of genetic information during subsequent cell division. hCAP-C/hCAP-E is a component of a multiprotein complex called condensin that interacts with phosphorylation of Histone H3 resulting in the mitotic chromosome condensation. The distribution patterns of the two heterodimeric complexes in interphase nucleus indicate independent behavior of the two complexes during cell cycle. These results suggest that the two distinct complexes are involved in different aspects of mitotic chromosome organization in humar cells.
TRUSTED_SUSTAINABILITY

Spezifikation

Antigen SMC2
Anwendungen ELISA, Western Blot
Klassifikation Polyclonal
Konzentration 0.66 mg/mL
Konjugat Unconjugated
Zusammensetzung PBS with 50% glycerol and 0.01% thimerosal; pH 7.3
Gen SMC2
Gen-Zugriffsnummer O95347, Q8CG48
Gen-Alias 5730502P04Rik; AI255214; AW545314; CAP E; CAPE; CAP-E; Chromosome-associated protein E; FGF-inducible protein 16; fibroblast growth factor inducible 16; Fin16; FLJ10093; hCAP-E; PRO0324; SMC protein 2; Smc2; SMC-2; SMC2 (structural maintenance of chromosomes 2, yeast)-like 1; SMC2 protein; SMC2 structural maintenance of chromosomes 2-like 1; Smc2l1; structural maintenance of chromosomes (SMC) family member, chromosome-associated protein E; structural maintenance of chromosomes 2; structural maintenance of chromosomes protein 2; XCAP-E homolog
Gensymbole SMC2
Wirtsspezies Rabbit
Immunogen Recombinant fusion protein containing a sequence corresponding to amino acids 700-900 of human SMC2 (NP_006435.2).
Reinigungsverfahren Affinity Chromatography
Menge 100 μL
Regulatorischer Status RUO
Primär oder sekundär Primary
Gen-ID (Entrez) 10592, 14211
Zielspezies Human, Mouse
Inhalt und Lagerung -20°C, Avoid Freeze/Thaw Cycles
Produkttyp Antibody
Form Liquid
Isotype IgG
Mehr anzeigen Weniger anzeigen
Name des Produkts
Wählen Sie ein Thema

Indem Sie auf Absenden klicken, erklären Sie sich damit einverstanden, dass Fisher Scientific sich mit Ihnen in Verbindung setzen kann, um Ihr Feedback in diesem Formular zu bearbeiten. Wir werden Ihre Informationen nicht für andere Zwecke weitergeben. Alle bereitgestellten Kontaktinformationen werden in Übereinstimmung mit unserer Datenschutzrichtlinie aufbewahrt. Datenschutzrichtlinie.