missing translation for 'onlineSavingsMsg'
Learn More
Learn More
SMARCAL1 Polyclonal Antibody, Invitrogen™
Rabbit Polyclonal Antibody
Marke: Invitrogen PA528980
Additional Details : Gewicht : 0.01000kg
Beschreibung
Recommended positive controls: A549, mouse testis, rat testis. Store product as a concentrated solution. Centrifuge briefly prior to opening the vial.
PRKCQ is a calcium-independent, novel isoform that is important in transducing signals in T lymphocytes. Interaction of a T lymphocyte with an antigen-presenting cell results in clustering of the T cell antigen receptor (TCR) and the recruitment of a large signaling complex to the site of cell-cell contact. PRKCQ is essential for TCR-mediated activation. PKC theta plays a role in pathways that link the TCR signaling complex to the activation of NF-kappa B in mature T lymphocytes.Spezifikation
SMARCAL1 | |
Polyclonal | |
Unconjugated | |
SMARCAL1 | |
6030401P21Rik; AI851716; ATP-driven annealing helicase; DNA-dependent ATPase A; HARP; HepA-related protein; HHARP; Mharp; SMARCAL1; SMARCA-like protein 1; Sucrose nonfermenting protein 2-like 1; Swi/SNF related matrix associated, actin dependent regulator of chromatin, subfamily a-like 1; SWI/SNF related, matrix associated, actin dependent regulator of chromatin, subfamily a like 1; SWI/SNF related, matrix associated, actin dependent regulator of chromatin, subfamily a-like 1; SWI/SNF-related matrix-associated actin-dependent regulator of chromatin subfamily A-like protein 1 | |
Rabbit | |
Antigen affinity chromatography | |
RUO | |
316477, 50485, 54380 | |
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
Liquid |
Immunocytochemistry, Immunoprecipitation, Western Blot | |
1.07 mg/mL | |
PBS with 20% glycerol and 0.025% ProClin 300; pH 7 | |
B4F769, Q8BJL0, Q9NZC9 | |
SMARCAL1 | |
Recombinant fragment corresponding to a region within amino acids 120 and 457 of Human SmarcAL1 | |
100 μL | |
Primary | |
Human, Mouse, Rat | |
Antibody | |
IgG |