missing translation for 'onlineSavingsMsg'
Learn More
Learn More
SLC9A10 Polyclonal Antibody, Invitrogen™
Rabbit Polyclonal Antibody
Marke: Invitrogen PA568333
Additional Details : Gewicht : 0.01000kg
Beschreibung
SLC9A10 is a member of the sodium-hydrogen exchanger (NHE) family (see SLC9A1, MIM 107310) and is required for male fertility and sperm motility (Wang et al., 2003 [PubMed 14634667]).Spezifikation
SLC9A10 | |
Polyclonal | |
Unconjugated | |
SLC9C1 | |
Gm610; LOW QUALITY PROTEIN: sodium/hydrogen exchanger 10; Na(+)/H(+) exchanger 10; NHE; NHE-10; Slc9a10; SLC9C1; sNHE; Sodium/hydrogen exchanger 10; solute carrier family 10 member 10; Solute carrier family 9 member 10; Solute carrier family 9 member C1; solute carrier family 9, isoform 10; solute carrier family 9, member 10; solute carrier family 9, subfamily C (Na+-transporting carboxylic acid decarboxylase), member 1; spermNHE; sperm-NHE; Sperm-specific Na(+)/H(+) exchanger; sperm-specific sodium proton exchanger | |
Rabbit | |
affinity chromatography | |
RUO | |
285335 | |
-20°C | |
Liquid |
Western Blot | |
1 mg/mL | |
PBS with 50% glycerol and 0.02% sodium azide; pH 7.4 | |
Q4G0N8 | |
SLC9C1 | |
Synthesized peptide derived from human SLC9A10. | |
100 μL | |
Primary | |
Human | |
Antibody | |
IgG |