missing translation for 'onlineSavingsMsg'
Learn More
Learn More
SLC38A10 Antibody, Novus Biologicals™
Rabbit Polyclonal Antibody
330.00 CHF - 643.00 CHF
Spezifikation
| Antigen | SLC38A10 |
|---|---|
| Verdünnung | Immunohistochemistry, Immunocytochemistry/Immunofluorescence 1 - 4 ug/ml, Immunohistochemistry-Paraffin 1:50 - 1:200 |
| Anwendungen | Immunohistochemistry, Immunocytochemistry, Immunofluorescence, Immunohistochemistry (Paraffin) |
| Klassifikation | Polyclonal |
| Konjugat | Unconjugated |
| Produktcode | Marke | Menge | Preis | Menge & Verfügbarkeit | |||||
|---|---|---|---|---|---|---|---|---|---|
| Produktcode | Marke | Menge | Preis | Menge & Verfügbarkeit | |||||
|
18400491
|
Novus Biologicals
NBP1-81195-25ul |
25ul |
330.00 CHF
25 Mikroliter |
Log in om dit product te kopen Registreer vandaag om een webaccount aan te maken | |||||
|
18297975
|
Novus Biologicals
NBP1-81195 |
0.1 mL |
643.00 CHF
0.10 Milliliter |
Log in om dit product te kopen Registreer vandaag om een webaccount aan te maken | |||||
Beschreibung
SLC38A10 Polyclonal specifically detects SLC38A10 in Human samples. It is validated for Immunohistochemistry, Immunocytochemistry/Immunofluorescence, Immunohistochemistry-Paraffin.Spezifikation
| SLC38A10 | |
| Immunohistochemistry, Immunocytochemistry, Immunofluorescence, Immunohistochemistry (Paraffin) | |
| Unconjugated | |
| RUO | |
| PBS (pH 7.2) and 40% Glycerol with 0.02% Sodium Azide | |
| 124565 | |
| This antibody was developed against Recombinant Protein corresponding to amino acids:PVPHDKVVVDEGQDREVPEENKPPSRHAGGKAPGVQGQMAPPLPDSEREKQEPEQGEVGKRPGQAQALEEAGDLPEDPQKVPEADGQPA | |
| Primary | |
| Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles. |
| Immunohistochemistry, Immunocytochemistry/Immunofluorescence 1 - 4 ug/ml, Immunohistochemistry-Paraffin 1:50 - 1:200 | |
| Polyclonal | |
| Rabbit | |
| Human | |
| FLJ35718, FLJ46399, MGC15523, PP1744, putative sodium-coupled neutral amino acid transporter 10, solute carrier family 38, member 10 | |
| SLC38A10 | |
| IgG | |
| Affinity Purified | |
| Specificity of antibody verified on a Protein Array containing target protein plus 383 other non-specific proteins. |
For Research Use Only
Haben Sie Verbesserungsvorschläge?Übermitteln Sie eine inhaltliche Korrektur
Berichtigung von Produktinhalten
Bitte geben Sie uns Ihr Feedback zu den Produktinhalten, indem Sie das folgende Formular ausfüllen.
Name des Produkts