missing translation for 'onlineSavingsMsg'
Learn More
Learn More
SLC37A4 Polyclonal Antibody, Invitrogen™
Rabbit Polyclonal Antibody
Marke: Thermo Scientific PA558600
Additional Details : Gewicht : 0.02000kg
Beschreibung
Immunogen sequence: NEPADVGLRN LDPMPSEGKK GSLKEESTLQ E Highest antigen sequence identity to the following orthologs: Mouse - 87%, Rat - 87%.
SLC37A4 transports glucose-6-phosphate from the cytoplasm to the lumen of the endoplasmic reticulum. It forms with glucose-6-phosphatase the complex responsible for glucose production through glycogenolysis and gluconeogenesis. Hence, it plays a central role in homeostatic regulation of blood glucose levels.Spezifikation
SLC37A4 | |
Polyclonal | |
Unconjugated | |
Slc37a4 | |
G6PT; G6PT1; G6PT2; G6PT3; glucose 6-phosphate transporter; glucose-5-phosphate transporter; glucose-6-phosphatase transport protein 1; glucose-6-phosphatase, transport (glucose) protein 3; glucose-6-phosphatase, transport (glucose-6-phosphate) protein 1; glucose-6-phosphatase, transport (phosphate/pyrophosphate) protein 2; glucose-6-phosphatase, transport protein 1; glucose-6-phosphate exchanger SLC37A4; Glucose-6-phosphate translocase; GSD1b; GSD-1b; GSD1c; GSD1d; mG6PT; MGC15729; microsomal glucose-6-phosphate transporter; PRO0685; SLC37A4; solute carrier family 37 (glucose-6-phosphate transporter), member 4; solute carrier family 37 (glycerol-6-phosphate transporter), member 4; solute carrier family 37 member 4; Transformation-related gene 19 protein; transporter; TRG19; TRG-19; Unknown (protein for MGC:137413) | |
Rabbit | |
Antigen affinity chromatography | |
RUO | |
2542 | |
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
Liquid |
Immunohistochemistry | |
0.8 mg/mL | |
PBS with 40% glycerol and 0.02% sodium azide; pH 7.2 | |
O43826 | |
Slc37a4 | |
Recombinant protein corresponding to Human SLC37A4 | |
100 μL | |
Primary | |
Human | |
Antibody | |
IgG |
Research Use Only