missing translation for 'onlineSavingsMsg'
Learn More

SLC25A25 Rabbit anti-Human, Mouse, Rat, Polyclonal, Proteintech

Rabbit Polyclonal Antibody

Marke:  Proteintech 21568-1-AP-150UL

Additional Details : Gewicht : 0.00010kg

 Lernen Sie weitere Varianten dieses

Artikelnummer. 16841165

  • 351.68 CHF / 150 Mikroliter

Verfügbar ab: 28-06-2022
Sie sich ein, um den Lagerbestand zu sehen
Zum Warenkorb hinzufügen

Beschreibung

Beschreibung

Calcium-dependent mitochondrial solute carrier. Mitochondrial solute carriers shuttle metabolites, nucleotides, and cofactors through the mitochondrial inner membrane. May act as a ATP-Mg/Pi exchanger that mediates the transport of Mg-ATP in exchange for phosphate, catalyzing the net uptake or efflux of adenine nucleotides into or from the mitochondria.
Spezifikation

Spezifikation

SLC25A25
Polyclonal
Unconjugated
SLC25A25
APC3, KIAA1896, MCSC, MCSC3, PCSCL, RP11 395P17.4, SCAMC 2, SCAMC2, SLC25A25
Rabbit
Antigen Affinity Chromatography
RUO
114789, 227731, 246771
Antibody
IgG
Immunohistochemistry (Paraffin), Western Blot
0.13 mg/mL
PBS with 50% glycerol and 0.02% sodium azide; pH 7.3
A2ASZ8, Q6KCM7, Q8K3P6
Slc25a25
SLC25A25 Fusion Protein Ag16086
150 μL
Primary
Human, Mouse, Rat
Liquid
Videos
Sicherheitsdatenblatt (SDS)
Dokumentation
Zertifikate
Sonderangebote

Sonderangebote