missing translation for 'onlineSavingsMsg'
Learn More
Learn More
SLC25A25 Rabbit anti-Human, Mouse, Rat, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Marke: Proteintech 21568-1-AP-20UL
Additional Details : Gewicht : 0.00010kg
Beschreibung
Calcium-dependent mitochondrial solute carrier. Mitochondrial solute carriers shuttle metabolites, nucleotides, and cofactors through the mitochondrial inner membrane. May act as a ATP-Mg/Pi exchanger that mediates the transport of Mg-ATP in exchange for phosphate, catalyzing the net uptake or efflux of adenine nucleotides into or from the mitochondria.Spezifikation
SLC25A25 | |
Polyclonal | |
Unconjugated | |
SLC25A25 | |
APC3, KIAA1896, MCSC, MCSC3, PCSCL, RP11 395P17.4, SCAMC 2, SCAMC2, SLC25A25 | |
Rabbit | |
Antigen Affinity Chromatography | |
RUO | |
114789, 227731, 246771 | |
-20°C | |
Liquid |
Immunohistochemistry (Paraffin), Western Blot | |
0.13 mg/mL | |
PBS with 50% glycerol and 0.02% sodium azide; pH 7.3 | |
A2ASZ8, Q6KCM7, Q8K3P6 | |
Slc25a25 | |
SLC25A25 Fusion Protein Ag16086 | |
20 μL | |
Primary | |
Rat, Human, Mouse | |
Antibody | |
IgG |