missing translation for 'onlineSavingsMsg'
Learn More
Learn More
SLC25A23 Rabbit anti-Human, Mouse, Rat, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Marke: Proteintech 20168-1-AP-20UL
Additional Details : Gewicht : 0.00010kg
Beschreibung
Calcium-dependent mitochondrial solute carrier. Mitochondrial solute carriers shuttle metabolites, nucleotides, and cofactors through the mitochondrial inner membrane. May act as a ATP-Mg/Pi exchanger that mediates the transport of Mg-ATP in exchange for phosphate, catalyzing the net uptake or efflux of adenine nucleotides into or from the mitochondria.Spezifikation
SLC25A23 | |
Polyclonal | |
Unconjugated | |
SLC25A23 | |
APC2, MCSC2, SCaMC 3, SCAMC3, SLC25A23 | |
Rabbit | |
Antigen Affinity Chromatography | |
RUO | |
301113, 66972, 79085 | |
Antibody | |
IgG |
Western Blot, Immunohistochemistry (Paraffin) | |
0.59 mg/mL | |
PBS with 50% glycerol and 0.02% sodium azide; pH 7.3 | |
Q6GQS1, Q9BV35 | |
Slc25a23 | |
SLC25A23 Fusion Protein Ag13987 | |
20 μL | |
Primary | |
Mouse, Human, Rat | |
Liquid |