missing translation for 'onlineSavingsMsg'
Learn More
Learn More
SLC1A4 Rabbit anti-Human, Mouse, Rat, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Marke: Proteintech 13067-2-AP-20UL
Additional Details : Gewicht : 0.00010kg
Beschreibung
Transporter for alanine, serine, cysteine, and threonine. Exhibits sodium dependence.Spezifikation
SLC1A4 | |
Polyclonal | |
Unconjugated | |
SLC1A4 | |
ASCT 1, ASCT1, SATT, SLC1A4 | |
Rabbit | |
Antigen Affinity Chromatography | |
RUO | |
305540, 55963, 6509 | |
Antibody | |
IgG |
Immunoprecipitation, Western Blot | |
0.41 mg/mL | |
PBS with 50% glycerol and 0.02% sodium azide; pH 7.3 | |
O35874, P43007 | |
Slc1a4 | |
SLC1A4 Fusion Protein Ag3763 | |
20 μL | |
Primary | |
Rat, Mouse, Human | |
Liquid |