missing translation for 'onlineSavingsMsg'
Learn More
Learn More
SIRT2 Rabbit anti-Human, Mouse, Rat, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Marke: Proteintech 19655-1-AP-150UL
Additional Details : Gewicht : 0.00010kg
Beschreibung
This antibody recognizes the 37-45 KD SIRT2 proteins. This antibody is a specific antibody that it can't detect signal with SIRT2-KO samples.
The SIRT2 protein (also known as Sirtuin for Silent Mating Type Information 2-Homolog) is a NAD-dependent deacytylase (NDAC) that has been shown to control gene silencing, cell cycle, and DNA damage repair. It is believed that SIRT2 may act as a tumor suppressor in human gliomas and may also serve as a novel molecular marker for these cells. SIRT2 has also been shown to act as a redox sensor to help regulate muscle gene expression in response to food intake and exercise. SIRT2 acts in the phosphorylation cascade involving mitosis where SIRT2 is phosphorylated in late G2 phase, during M phase, and into cytokinesis.Spezifikation
SIRT2 | |
Polyclonal | |
Unconjugated | |
SIRT2 | |
SIR2, SIR2 like protein 2, SIR2L, SIR2L2, SIRT2 | |
Rabbit | |
Antigen Affinity Chromatography | |
RUO | |
22933, 361532, 64383 | |
-20°C | |
Liquid |
Immunofluorescence, Immunohistochemistry (Paraffin), Immunoprecipitation, Western Blot | |
0.29 mg/mL | |
PBS with 50% glycerol and 0.1% sodium azide; pH 7.3 | |
Q5RJQ4, Q8IXJ6, Q8VDQ8 | |
Sirt2 | |
SIRT2-Specific Fusion Protein Ag7756 | |
150 μL | |
Primary | |
Human, Mouse, Rat | |
Antibody | |
IgG |