Learn More
Thermo Scientific™ Sino Biological™ TREM2 Recombinant Human Protein, His Tag
This recombinant human protein is expressed from a DNA sequence encoding the extracellular domain of human TREM2 (NP_061838.1) (Met 1-Ser 174) fused to a polyhistidine tag at the C-terminus
Marke: Thermo Scientific™ 11084H08H5
Additional Details : Gewicht : 0.01000kg
Beschreibung
Triggering Receptor Expressed on Myeloid Cells 2 (TREM2) recombinant human protein is supplied as a lyophilized powder. This protein is suitable for use in analysis of protein structure. In general, recombinant proteins can also be used as an immunogen, as a protein standard, or in cell biology research applications.&line-break;Background: Triggering receptor expressed on myeloid cells 2 (TREM2) is a single Ig domain receptor. It is expressed on macrophages and dendritic cells but not on granulocytes or monocytes. TREM2 may play a role in chronic inflammations and may stimulate production of constitutive rather than inflammatory chemokines and cytokines. TREM2 forms a receptor signaling complex with TYROBP and triggers activation of the immune responses in macrophages and dendritic cells. It also associates with the signal adapter protein, DAP12, which has a cytoplasmic ITAM, leading to the subsequent activation of cytoplasmic tyrosine kinases. TREM2 is both required and sufficient for competent uptake of apoptotic neuronal cells. TREM2 and TREM2-L form a receptor-ligand pair connecting microglia with apoptotic neurons, directing removal of damaged cells to allow repair. Deficiency of the adapter protein DAP12 or its associated receptor TREM2 is associated with abnormal osteoclast development in humans. Defects in TREM2 are causes of PLOSL, also known as NHD. In addition, TREM2 signaling is also an important pathway to promote healing.- N-terminal Sequence Analysis: His 19
- Activity: The activity of this protein has not been tested
- Formulation: lyophilized in PBS, pH 7.4, 5% mannitol, 5% trehalose, 0.02% Tween™-80
- Reconstitution: dissolve the protein in sterile double-distilled water to a concentration of 0.2mg/mL or lower; aliquot and use as soon as possible
Manufactured by Sino Biological (China).
Spezifikation
Protein Expression and Isolation, Enzyme and Protein Activity Assay, Protein Assay Control, Reference Standards and Accessories | |
18.9kDa | |
5μg | |
C-Terminal | |
<1 EU/μg | |
Triggering receptor expressed on myeloid cells 2 | |
His Tag | |
Other Proteins | |
Other Proteins |
54209 | |
PAGE | |
Store at -20°C. Store aliquots under sterile conditions at -20°C. Avoid repeated freeze-thaw cycles. Expires one year from date of receipt when stored as instructed. | |
Truncated/Recombinant | |
TREM-2, Trem2a, Trem2b, Trem2c, TREM2 | |
TREM2 | |
Mammalian | |
Lyophilized | |
≥85% by SDS-PAGE |
For Research Use Only. Not for use in diagnostic procedures.