missing translation for 'onlineSavingsMsg'
Learn More

PARP1 Recombinant Human Protein, His Tag, Invitrogen™ Sino Biological™

This recombinant protein was expressed from the amino acids corresponding to the full length of human PARP1 (NP_001609.2) (Met 1-Trp 1014) fused to a polyhistidine tag at the C-terminus

Marke:  Invitrogen 11040H08B50

Additional Details : Gewicht : 0.10000kg

 Lernen Sie weitere Varianten dieses

Artikelnummer. 15267615

Dieser Artikel wurde eingestellt und ist leider nicht mehr verfügbar. Sehen Sie sich das Produkt an, um alternative Produktvorschläge zu sehen oder kontaktieren Sie unseren Technischen Support unter 056 618 41 11.


Beschreibung

Beschreibung

Poly (Adp-Ribose) Polymerase 1 (PARP1), recombinant human protein is supplied in frozen format. It is suitable for use in protein studies such as protein structure analysis and protein-protein interactions. In general, recombinant proteins can also be used as an immunogen, as a protein standard, or in cell biology research applications.
  • Activity: Measured by its binding ability in a functional ELISA; immobilized human PARP1 at 10μg/mL (100μL/well) can bind to biotinylated human HSP70, the EC50 of biotinylated human HSP70 is 0.035μg/mL
  • Formulation: 50mM Tris, 100mM NaCl, pH 8, 2mM EDTA, 2mM PMSF, 0.5mM TCEP, 10% glycerol
  • Reconstitution: No reconstitution needed

This protein is manufactured by Sino Biological Inc.

Spezifikation

Spezifikation

Protein Expression and Isolation, Enzyme and Protein Activity Assay, Protein Assay Control, Reference Standards and Accessories
50μg
<1 EU/μg
Poly (ADP-ribose) polymerase 1
His Tag
Frozen
≥90% by SDS-PAGE
114.5kDa
Store at -80°C. Avoid repeated freeze-thaw cycles. Expires one year from date of receipt when stored as instructed.
PARP1, RP11-125A15.2, ADPRT, ADPRT1, PARP, PARP-1, PPOL, pADPRT-1
PARP1
Insect
Other Proteins
Produktvorschläge

Produktvorschläge

Videos
Sicherheitsdatenblatt (SDS)
Dokumentation

Dokumentation

Zertifikate
Sonderangebote

Sonderangebote

For Research Use Only. Not for use in diagnostic procedures.