missing translation for 'onlineSavingsMsg'
Learn More

NRG1 Recombinant Human Protein, His Tag, Invitrogen™ Sino Biological™

This recombinant protein was expressed from a DNA sequence encoding the extracellular domain of human NRG1 isoform alpha (Q02297-1) (Ser 20-Lys 241) fused to the Fc region of human IgG1 at the C-terminus

Marke:  Invitrogen 13499H08H5

Additional Details : Gewicht : 0.01000kg

Artikelnummer. 15236325

Dieser Artikel wurde eingestellt und ist leider nicht mehr verfügbar. Sehen Sie sich das Produkt an, um alternative Produktvorschläge zu sehen oder kontaktieren Sie unseren Technischen Support unter 056 618 41 11.


Beschreibung

Beschreibung

Neuregulin 1 (NRG1), recombinant human protein is supplied as a lyophilized powder. It is suitable for use in protein studies such as protein structure analysis and protein-protein interactions. In general, recombinant proteins can also be used as an immunogen, as a protein standard, or in cell biology research applications.
  • Activity: This recombinant protein has not been tested
  • Formulation: Lyophilized in 140mM NaCl, 2.7mM KCl, 10mM Na2HPO4, 1.8mM KH2PO4, pH 7.4, 5% Mannitol, 5% Trehalose, 0.02% Tween 80
  • Reconstitution: Dissolve the protein in sterile double distilled water to a concentration of 0.2mg/mL or lower; aliquot and use as soon as possible

This protein is manufactured by Sino Biological Inc.

Spezifikation

Spezifikation

Protein Expression and Isolation, Enzyme and Protein Activity Assay, Protein Assay Control, Reference Standards and Accessories
5μg
<1 EU/μg
Neuregulin 1
His Tag
Growth Factors & Receptors
Other Growth Factors
25.5kDa
Store at -20°C. Store aliquots under sterile conditions at -20°C. Avoid repeated freeze-thaw cycles. Expires one year from date of receipt when stored as instructed.
ARIA, GGF, GGF2, HGL, HRG, HRG1, HRGA, MST131, NDF, SMDF
NRG1
Mammalian
Lyophilized
≥95% by SDS-PAGE
Produktvorschläge

Produktvorschläge

Videos
Sicherheitsdatenblatt (SDS)
Dokumentation

Dokumentation

Zertifikate
Sonderangebote

Sonderangebote

For Research Use Only. Not for use in diagnostic procedures.