missing translation for 'onlineSavingsMsg'
Learn More

IL13 Recombinant Cynomolgus Protein, N-His Tag, Invitrogen™ Sino Biological™

This recombinant protein was expressed from a DNA sequence encoding the mature form of cynomolgus (Macaca fascicularis) IL-13 (ABG75889.1) (Ser 21-Asn 132) fused to a polyhistidine tag at the N-terminus

Marke:  Invitrogen 11057C07H25

Additional Details : Gewicht : 0.10000kg

Artikelnummer. 15217665

Dieser Artikel wurde eingestellt und ist leider nicht mehr verfügbar. Sehen Sie sich das Produkt an, um alternative Produktvorschläge zu sehen oder kontaktieren Sie unseren Technischen Support unter 056 618 41 11.


Beschreibung

Beschreibung

Interleukin 13 (IL13), recombinant cynomolgus protein is supplied as a lyophilized powder. It is suitable for use in performing cell-based assays and protein studies such as protein structure analysis and protein-protein interactions. In general, recombinant proteins can also be used as an immunogen, as a protein standard, or in cell biology research applications.
  • Activity: 1. Measured by its binding ability in a functional ELISA; immobilized cynomolgus IL-13 at 4μg/mL (100μL/well) can bind human IL13RA1 with a linear ranger of 0.156-2.5μg/mL; 2. Measured in a cell proliferation assay using TF-1 human erythroleukemic cells, the specific activity is 5-20ng/mL
  • Formulation: Lyophilized in 140mM NaCl, 2.7mM KCl, 10mM Na2HPO4, 1.8mM KH2PO4, pH 7.4, 5% Mannitol, 5% Trehalose, 0.02% Tween 80
  • Reconstitution: Dissolve the protein in sterile double distilled water to a concentration of 0.2mg/mL or lower; aliquot and use as soon as possible

This protein is manufactured by Sino Biological Inc.

Spezifikation

Spezifikation

Protein Expression and Isolation, Enzyme and Protein Activity Assay, Protein Assay Control, Reference Standards and Accessories
5 x 5μg
<1 EU/μg
Interleukin 13
N-His Tag
CD Proteins
Interleukins
14.5kDa
Store at -20°C. Store aliquots under sterile conditions at -20°C. Avoid repeated freeze-thaw cycles. Expires one year from date of receipt when stored as instructed.
IL-13
IL13
Mammalian
Lyophilized
≥90% by SDS-PAGE
Produktvorschläge

Produktvorschläge

Videos
Sicherheitsdatenblatt (SDS)
Dokumentation

Dokumentation

Zertifikate
Sonderangebote

Sonderangebote

For Research Use Only. Not for use in diagnostic procedures.