missing translation for 'onlineSavingsMsg'
Learn More

APOA1 (APOAI) Recombinant Human Protein, hIgG1-Fc Tag, Invitrogen™ Sino Biological™

This recombinant protein was expressed from a DNA sequence encoding the pro form of human APOA1 (CAA26097.1) (Met 1-Gln 267) was fused with Fc region of human IgG1 at the C-terminus

Marke:  Invitrogen 10686H02H5

Additional Details : Gewicht : 0.10000kg

Artikelnummer. 15267045

Dieser Artikel wurde eingestellt und ist leider nicht mehr verfügbar. Sehen Sie sich das Produkt an, um alternative Produktvorschläge zu sehen oder kontaktieren Sie unseren Technischen Support unter 056 618 41 11.


Beschreibung

Beschreibung

Apolipoprotein A-I (APOAI), recombinant human protein is supplied as a lyophilized powder. It is suitable for use in protein studies such as protein structure analysis and protein-protein interactions. In general, recombinant proteins can also be used as an immunogen, as a protein standard, or in cell biology research applications.
  • Activity: Measured by its ability to inhibit proliferation of HeLa human cervical epithelial carcinoma cells, the ED50 for this effect is typically 2.5-10μg/mL
  • Formulation: Lyophilized in 140mM NaCl, 2.7mM KCl, 10mM Na2HPO4, 1.8mM KH2PO4, pH 7.4, 5% Mannitol, 5% Trehalose, 0.02% Tween 80
  • Reconstitution: Dissolve the protein in sterile double distilled water to a concentration of 0.2mg/mL or lower; aliquot and use as soon as possible

This protein is manufactured by Sino Biological Inc.

Spezifikation

Spezifikation

Protein Expression and Isolation, Enzyme and Protein Activity Assay, Protein Assay Control, Reference Standards and Accessories
5μg
<1 EU/μg
Apolipoprotein A-I
IgG1 Fc Tag
Other Proteins
Other Proteins
55kDa
Store at -20°C. Store aliquots under sterile conditions at -20°C. Avoid repeated freeze-thaw cycles. Expires one year from date of receipt when stored as instructed.
APOA1, mgC117399
APOA1
Mammalian
Lyophilized
≥90% by SDS-PAGE
Produktvorschläge

Produktvorschläge

Videos
Sicherheitsdatenblatt (SDS)
Dokumentation

Dokumentation

Zertifikate
Sonderangebote

Sonderangebote

For Research Use Only. Not for use in diagnostic procedures.