missing translation for 'onlineSavingsMsg'
Learn More

AMICA1 (JAML) Recombinant Human Protein, His Tag, Invitrogen™ Sino Biological™

This recombinant protein was expressed from a DNA sequence encoding the extracellular domain of human AMICA1 (NP_001091996.1) (Met 1-Leu 275) fused to a polyhistidine tag at the C-terminus

Marke:  Invitrogen 10120H08H5

Additional Details : Gewicht : 0.10000kg

Artikelnummer. 15234685

Dieser Artikel wurde eingestellt und ist leider nicht mehr verfügbar. Sehen Sie sich das Produkt an, um alternative Produktvorschläge zu sehen oder kontaktieren Sie unseren Technischen Support unter 056 618 41 11.


Beschreibung

Beschreibung

Adhesion molecule, interacts with CXADR antigen 1 (AMICA1), recombinant human protein is supplied as a lyophilized powder. It is suitable for use in analysis of protein structure, and performing cell based assays. It can also be used as an immunogen, as a protein standard, or in other research applications.
  • Activity: Measured by its ability to inhibit NGF-induced proliferation of TF1 human erythroleukemic cells
  • Formulation: Lyophilized in 140mM NaCl, 2.7mM KCl, 10mM Na2HPO4, 1.8mM KH2PO4, pH 7.4, 5% Mannitol, 5% Trehalose, 0.02% Tween 80
  • Reconstitution: Dissolve the protein in sterile double distilled water to a concentration of 0.2mg/mL or lower; aliquot and use as soon as possible

This protein is manufactured by Sino Biological Inc.

Spezifikation

Spezifikation

Protein Expression and Isolation, Enzyme and Protein Activity Assay, Protein Assay Control, Reference Standards and Accessories
5μg
<1 EU/μg
Adhesion molecule, interacts with CXADR antigen 1
His Tag
Other Proteins
Intercellular Adhesion Molecule
30.5kDa
Store at -20°C. Store aliquots under sterile conditions at -20°C. Avoid repeated freeze-thaw cycles. Expires one year from date of receipt when stored as instructed.
JAML, AMICA, Gm638, CREA7-1, CREA7-4, FLJ37080, mgC118814, MGC118815, AMICA1
AMICA1
Mammalian
Lyophilized
≥85% by SDS-PAGE
Produktvorschläge

Produktvorschläge

Videos
Sicherheitsdatenblatt (SDS)
Dokumentation

Dokumentation

Zertifikate
Sonderangebote

Sonderangebote

For Research Use Only. Not for use in diagnostic procedures.