Learn More
Cole-Parmer™ X25 MEMBRANE 0.2UM 47MM
Marke: Cole-Parmer™ 06741-14
Additional Details : Gewicht : 0.16000kg
Spezifikation
350L/cm2 | |
Silver | |
25Pack | |
47 mm |
13psi | |
0,2 μm | |
Filter |