missing translation for 'onlineSavingsMsg'
Learn More

Siliconöl, hochsiedend, Thermo Scientific Chemicals

Catalog Number p-4323797
Click to view available options
Menge:
100 g
500 g
2.5 kg
Verpackung:
Glasflasche
63148-58-3
Poly(methylphenylsiloxane)
63148-58-3
Poly(methylphenylsiloxane)

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY
Chemischer Name oder Material Silicone oil
Dichte 1.0500g/mL
Flammpunkt 316°C
Verpackung Glasflasche
Brechungsindex 1.4940 to 1.5010
Menge 100 g
Relative Dichte 1.05
Löslichkeitsinformationen Solubility in water: insoluble in water.
Viskosität 125 mm2/s (25°C)

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'