Learn More
Elma™ Silicone Mat for Type 300/900
This silicone mat is to be placed into the basket for sensitive parts protection.
Marke: Elma™ 1117991
Additional Details : Gewicht : 0.10000kg
Beschreibung
- Must be cut accordingly for the different basket sizes
- Lets you clearly structure your cleaning process
- Prevents scratches on the tank floor
Spezifikation
Mat | |
Fisherbrand Select, Fisherbrand EASY, Elmasonic P, Elmasonic Select, Elmasonic EASY | |
Silicone |