Learn More
Endecotts™ Stainless Steel Test Sieve, 450mm dia.
Full height test sieves for all types of laboratory sampling and particle size analysis. Endecotts™ Stainless Steel Test Sieve, 450mm dia. comply with ISO 3310-1 / BS410 standards.
Marke: Endecotts™ 450SIW20.0
Additional Details : Gewicht : 0.10000kg
Beschreibung
- Stainless steel precision frame (ensures consistent nestability)
- Stainless steel mesh
- Precise aperture in accordance with ISO 3310-1 / BS410
- Natural fillet (free flowing of sample)
- Totally sealed (no crevice to lose material)
- Evenly tensioned mesh (ensures accurate analysis)
- Safe edge (big radius makes it comfortable to handle)
- Serial number (ensures full traceability)