Learn More
Sharpie EIGHT-COLOR PERMANENT MARKER ASSORTMENT SET OF 8
This Eight-color Permanent Marker Assortment Set offers pen-style, all-purpose markers with fine (1 mm) tips.
Marke: Sharpie 09968-80
47.55 CHF gültig bis 2024-04-30
Benutzen Sie den Promotions-Code "21673" um Ihren Promotions-Preis zu erhalten.
Additional Details : Gewicht : 0.09000kg
Beschreibung
- Quick drying, water-resistant ink in eight brilliant colors.
- Certified nontoxic; conform to ASTM D-4236.
- Assortment sets are available separately.
Spezifikation
Permanentmarkersatz | |
Assorted | |
8 Packungen |
Fine | |
8 Marker | |
1 mm |