Learn More
Abnova™ SGCD Recombinant Protein
Marke:  Abnova™ H00006444-P01.25ug
Additional Details : Gewicht : 0.00010kg
 Lernen Sie weitere Varianten dieses
Artikelnummer. 16130712
  • 547.00 CHF / 25 Mikrogramm
Verfügbar ab: 11-06-2024
Sie sich ein, um den Lagerbestand zu sehen
Zum Warenkorb hinzufügen
Beschreibung
Beschreibung
ELISA, Western Blotting (Recombinant Protein), Antibody Production, Protein Array
Spezifikation
Spezifikation
AAH20740
50mM Tris HCl, 10mM reduced Glutathione, pH 8 in the Elution Buffer
57.42
8
Glutathione Sepharose 4 Fast Flow
25 ug
MMPQEQYTHHRSTMPGSVGPQVYKVGIYGWRKRCLYFFVLLLMILILVNLAMTIWILKVMNFTIDGMGNLRITEKGLKLEGDSEFLQPLYAKEIQSRPGNALYFKSARNVTVNILNDQTKVLTQLITGPKAVEAYGKKFEVKTVSGKLLFSADNNEVVVGAERLRVLGAEGTVFPKSIETPNVRADPFKELRLESPTRSLVMEAPKGVEINAEAGNMEATCRTELRLESKDGEIKLDAAKIRLPRLPHGSYTPTGTRQKVFEICVCANGRLFLSQAGAGSTCQINTSVCL
35DAG/CMD1L/DAGD/MGC22567/SG-delta/SGCDP/SGD
SGCD
Wheat Germ (in vitro)
GST
Antibody Production, Protein Array, ELISA, Western Blot
6444
SGCD (Human) Recombinant Protein (P01)
In vitro wheat germ expression system
12.5% SDS-PAGE Stained with Coomassie Blue.
Wheat Germ (in vitro)
Store at -80°C. Aliquot to avoid repeated freezing and thawing.
SGCD
Human
Recombinant
Solution