Learn More
SFTPC Polyclonal Antibody, Invitrogen™
Rabbit Polyclonal Antibody
Marke: Thermo Scientific PA552759
Additional Details : Gewicht : 0.02000kg
Beschreibung
Immunogen sequence: PEAQQRLALS EHLVTTATFS IGSTGLVVYD YQQLLIAYKP APGTCCYIMK IAPESIPSLE ALTRKVHNFQ AKPAVPTSKL GQAEGRDAGS APSGGDPAFL GMAVSTLCG Highest antigen sequence identity to the following orthologs: Mouse - 75%, Rat - 76%.
This gene encodes the pulmonary-associated surfactant protein C (SPC), an extremely hydrophobic surfactant protein essential for lung function and homeostasis after birth. Pulmonary surfactant is a surface-active lipoprotein complex composed of 90% lipids and 10% proteins which include plasma proteins and apolipoproteins SPA, SPB, SPC and SPD. The surfactant is secreted by the alveolar cells of the lung and maintains the stability of pulmonary tissue by reducing the surface tension of fluids that coat the lung. Multiple mutations in this gene have been identified, which cause pulmonary surfactant metabolism dysfunction type 2, also called pulmonary alveolar proteinosis due to surfactant protein C deficiency, and are associated with interstitial lung disease in older infants, children, and adults. Alternatively spliced transcript variants encoding different protein isoforms have been identified.Spezifikation
SFTPC | |
Polyclonal | |
Unconjugated | |
Sftpc | |
BRICD6; BRICHOS domain containing 6; LOW QUALITY PROTEIN: pulmonary surfactant-associated protein C; pro-SpC; PSP C; PSP-C; pulmonary surfactant apoprotein-2 SP-C; pulmonary surfactant associated protein C; pulmonary surfactant protein C; pulmonary surfactant-associated protein C; Pulmonary surfactant-associated proteolipid SPL(Val); SFTP 2; SFTP2; Sftp-2; Sftpc; SMDP2; SP C; SP5; SPC; SP-C; surfactant associated protein C; surfactant protein C; Surfactant pulmonary-associated protein C; Surfactant, pulmonary-associated protein C; surfactant-associated protein; type I SP-C; type I surfactant protein C; type II SP-C; type II surfactant protein C | |
Rabbit | |
Antigen affinity chromatography | |
RUO | |
6440 | |
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
Liquid |
Immunohistochemistry, Western Blot | |
0.25 mg/mL | |
PBS with 40% glycerol and 0.02% sodium azide; pH 7.2 | |
P11686 | |
Sftpc | |
Recombinant protein corresponding to Human SFTPC | |
100 μL | |
Primary | |
Human | |
Antibody | |
IgG |
Research Use Only