missing translation for 'onlineSavingsMsg'
Learn More
Learn More
SFT2D3 Rabbit anti-Human, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Marke: Proteintech 19758-1-AP-20UL
Additional Details : Gewicht : 0.00010kg
Beschreibung
SFT2D3 may be involved in fusion of retrograde transport vesicles derived from an endocytic compartment with the Golgi complex.Spezifikation
SFT2D3 | |
Polyclonal | |
Unconjugated | |
SFT2D3 | |
SFT2 domain containing 3, SFT2D3 | |
Rabbit | |
Antigen Affinity Chromatography | |
RUO | |
84826 | |
-20°C | |
Liquid |
Western Blot, Immunohistochemistry (Paraffin) | |
0.13 mg/mL | |
PBS with 50% glycerol and 0.02% sodium azide; pH 7.3 | |
Q587I9 | |
SFT2D3 | |
Peptide | |
20 μL | |
Primary | |
Human | |
Antibody | |
IgG |