missing translation for 'onlineSavingsMsg'
Learn More
Learn More
SETD7 Rabbit anti-Human, Mouse, Rat, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Marke: Proteintech 24840-1-AP-20UL
Dieser Artikel kann nicht zurückgegeben werden.
Rückgaberichtlinie anzeigen
Beschreibung
Histone methyltransferase that specifically monomethylates ′Lys-4′ of histone H3. H3 ′Lys-4′ methylation represents a specific tag for epigenetic transcriptional activation. Plays a central role in the transcriptional activation of genes such as collagenase or insulin. Recruited by IPF1/PDX-1 to the insulin promoter, leading to activate transcription. Has also methyltransferase activity toward non-histone proteins such as p53/TP53, TAF10, and possibly TAF7 by recognizing and binding the [KR]-[STA]-K in substrate proteins. Monomethylates ′Lys-189′ of TAF10, leading to increase the affinity of TAF10 for RNA polymerase II. Monomethylates ′Lys-372′ of p53/TP53, stabilizing p53/TP53 and increasing p53/TP53-mediated transcriptional activation. Also able to demethylated ′Lys-372′ of p53/TP53 in vitro.Spezifikation
| SETD7 | |
| Polyclonal | |
| Unconjugated | |
| SETD7 | |
| H3 K4 HMTase SETD7, KIAA1717, KMT7, Lysine N methyltransferase 7, SET7, SET7/9, SET9, SETD7 | |
| Rabbit | |
| Antigen Affinity Chromatography | |
| RUO | |
| 689954, 73251, 80854 | |
| -20°C | |
| Liquid |
| Immunofluorescence, Western Blot, Immunocytochemistry | |
| 0.41 mg/mL | |
| PBS with 50% glycerol and 0.1% sodium azide; pH 7.3 | |
| Q8VHL1, Q8WTS6 | |
| Setd7 | |
| SETD7 Fusion Protein Ag18219 | |
| 20 μL | |
| Primary | |
| Human, Mouse, Rat | |
| Antibody | |
| IgG |
Berichtigung von Produktinhalten
Bitte geben Sie uns Ihr Feedback zu den Produktinhalten, indem Sie das folgende Formular ausfüllen.
Name des Produkts
Haben Sie Verbesserungsvorschläge?Übermitteln Sie eine inhaltliche Korrektur