missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Serpin C1/Antithrombin-III Antibody, Novus Biologicals™
Rabbit Polyclonal Antibody has been used in 2 publications
418.00 CHF - 595.00 CHF
Spezifikation
| Antigen | Serpin C1/Antithrombin-III |
|---|---|
| Konzentration | 0.1mg/mL |
| Verdünnung | Immunohistochemistry, Immunohistochemistry-Paraffin 1:50 - 1:200 |
| Anwendungen | Immunohistochemistry, Immunohistochemistry (Paraffin) |
| Klassifikation | Polyclonal |
| Produktcode | Marke | Menge | Preis | Menge & Verfügbarkeit | |||||
|---|---|---|---|---|---|---|---|---|---|
| Produktcode | Marke | Menge | Preis | Menge & Verfügbarkeit | |||||
|
18471202
|
Novus Biologicals
NBP1-90076-25ul |
25 μL |
418.00 CHF
25 Mikroliter |
Log in om dit product te kopen Registreer vandaag om een webaccount aan te maken | |||||
|
18276248
|
Novus Biologicals
NBP1-90076 |
0.1 mL |
595.00 CHF
0.10 Milliliter |
Log in om dit product te kopen Registreer vandaag om een webaccount aan te maken | |||||
Beschreibung
Serpin C1/Antithrombin-III Polyclonal specifically detects Serpin C1/Antithrombin-III in Human samples. It is validated for Immunohistochemistry, Immunohistochemistry-Paraffin.Spezifikation
| Serpin C1/Antithrombin-III | |
| Immunohistochemistry, Immunohistochemistry-Paraffin 1:50 - 1:200 | |
| Polyclonal | |
| Rabbit | |
| Human | |
| AT3antithrombin-III, ATIIIantithrombin III, MGC22579, serine (or cysteine) proteinase inhibitor, clade C (antithrombin), member 1, serine-cysteine proteinase inhibitor clade C member 1, Serpin C1, serpin peptidase inhibitor, clade C (antithrombin), member 1 | |
| SERPINC1 | |
| IgG | |
| Affinity Purified | |
| Specificity of antibody verified on a Protein Array containing target protein plus 383 other non-specific proteins. |
| 0.1mg/mL | |
| Immunohistochemistry, Immunohistochemistry (Paraffin) | |
| Unconjugated | |
| RUO | |
| PBS (pH 7.2) and 40% Glycerol with 0.02% Sodium Azide | |
| 462 | |
| This antibody was developed against Recombinant Protein corresponding to amino acids:HGSPVDICTAKPRDIPMNPMCIYRSPEKKATEDEGSEQKIPEATNRRVWELSKANSRFATTFYQHLADSKNDNDNIFLSPLSISTAFAMTKLGACNDTLQQLMEVFKFDTISEKTSDQIHFFFAKLNCRLYR | |
| Primary | |
| Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles. |
For Research Use Only
Haben Sie Verbesserungsvorschläge?Übermitteln Sie eine inhaltliche Korrektur
Berichtigung von Produktinhalten
Bitte geben Sie uns Ihr Feedback zu den Produktinhalten, indem Sie das folgende Formular ausfüllen.
Name des Produkts