missing translation for 'onlineSavingsMsg'
Learn More
Learn More
SERINC2 Rabbit anti-Human, Mouse, Rat, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
133.80 CHF - 381.34 CHF
Spezifikation
Antigen | SERINC2 |
---|---|
Konzentration | 0.13 mg/mL |
Anwendungen | Western Blot |
Klassifikation | Polyclonal |
Konjugat | Unconjugated |
Produktcode | Marke | Menge | Preis | Menge & Verfügbarkeit | |||||
---|---|---|---|---|---|---|---|---|---|
Produktcode | Marke | Menge | Preis | Menge & Verfügbarkeit | |||||
16859794
|
Proteintech
20266-1-AP-20UL |
20 μL |
133.80 CHF
20 Mikroliter |
Verfügbar ab: 05-06-2024
Loggen Sie sich ein, um den Lagerbestand zu sehen |
|||||
16849794
|
Proteintech
20266-1-AP-150UL |
150 μL |
381.34 CHF
150 Mikroliter |
Verfügbar ab: 05-06-2024
Loggen Sie sich ein, um den Lagerbestand zu sehen |
|||||
Beschreibung
The antibody is specific to SERINC2.
This magnesium-dependent enzyme catalyzes the hydrolysis of ATP coupled with the translocation of calcium from the cytosol to the sarcoplasmic reticulum lumen. Isoform 2 is involved in the regulation of the contraction/relaxation cycle. Wang Y. -G., Submitted (FEB-2001) to the EMBL/GenBank/DDBJ databases. Clark H. F., Genome Res. 13:2265-2270(2003). The MGC Project Team, Genome Res. 14:2121-2127(2004).Spezifikation
SERINC2 | |
Western Blot | |
Unconjugated | |
Rabbit | |
Human, Mouse, Rat | |
Q8K0E7, Q96SA4 | |
230779, 313057, 347735 | |
Peptide | |
Primary | |
-20°C | |
SERINC2 |
0.13 mg/mL | |
Polyclonal | |
Liquid | |
RUO | |
PBS with 50% glycerol and 0.02% sodium azide; pH 7.3 | |
FKSG84, PRO0899, SERINC2, serine incorporator 2, TDE2, TDE2L, UNQ263/PRO300 | |
Serinc2 | |
IgG | |
Antigen Affinity Chromatography | |
Antibody |