missing translation for 'onlineSavingsMsg'
Learn More
Learn More
SEPT8 Rabbit anti-Human, Mouse, Rat, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Marke: Proteintech 11769-1-AP-150UL
Additional Details : Gewicht : 0.00010kg
Beschreibung
SEPT8 is a member of the highly conserved septin family. Septins are 40- to 60-kDa GTPases that assemble as filamentous scaffolds. They are involved in the organization of submembranous structures, in neuronal polarity, and in vesicle trafficking (Blaser et al., 2003 ).Spezifikation
SEPT8 | |
Polyclonal | |
Unconjugated | |
SEPT8 | |
KIAA0202, SEP2, SEPT8, septin 8 | |
Rabbit | |
Antigen Affinity Chromatography | |
RUO | |
20362, 23176, 303135 | |
-20°C | |
Liquid |
Immunohistochemistry (Paraffin), Immunoprecipitation, Western Blot | |
0.3 mg/mL | |
PBS with 50% glycerol and 0.1% sodium azide; pH 7.3 | |
B0BNF1, Q8CHH9, Q92599 | |
SEPT8 | |
SEPT8 Fusion Protein Ag2357 | |
150 μL | |
Primary | |
Human, Mouse, Rat | |
Antibody | |
IgG |