missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ Selenoprotein W Polyclonal Antibody

Rabbit Polyclonal Antibody
Marke: Invitrogen™ PA567614
Dieser Artikel kann nicht zurückgegeben werden.
Rückgaberichtlinie anzeigen
Beschreibung
Selenoprotein W Polyclonal Antibody for IHC (P)
This gene encodes a selenoprotein containing a selenocysteine (Sec) residue, which is encoded by the UGA codon that normally signals translation termination. The 3' UTRs of selenoprotein mRNAs contain a conserved stem-loop structure, the Sec insertion sequence (SECIS) element that is necessary for the recognition of UGA as a Sec codon rather than as a stop signal. This protein is highly expressed in skeletal muscle, heart and brain. It belongs to the SelWTH family, which possesses a thioredoxin-like fold and a conserved CxxU (C is cysteine, U is Sec) motif, suggesting a redox function for this gene. Studies in mouse show that this selenoprotein is involved in muscle growth and differentiation, and in the protection of neurons from oxidative stress during neuronal development. A retroprocessed pseudogene for this locus has been identified on chromosome 1.
Spezifikation
| Selenoprotein W | |
| Polyclonal | |
| Unconjugated | |
| SELENOW | |
| Selenoprotein W; selenoprotein W, 1; selenoprotein W, muscle 1; Selenow; SelW; Sepw1 | |
| Rabbit | |
| Antigen affinity chromatography | |
| RUO | |
| 6415 | |
| -20°C | |
| Liquid |
| Immunohistochemistry (Paraffin) | |
| 0.8 mg/mL | |
| PBS with 40% glycerol and 0.05% sodium azide; pH 7.4 | |
| P63302 | |
| SELENOW | |
| Synthesized peptide derived from human Selenoprotein W. | |
| 100 μL | |
| Primary | |
| Human | |
| Antibody | |
| IgG |
Berichtigung von Produktinhalten
Bitte geben Sie uns Ihr Feedback zu den Produktinhalten, indem Sie das folgende Formular ausfüllen.
Name des Produkts
Haben Sie Verbesserungsvorschläge?Übermitteln Sie eine inhaltliche Korrektur