Learn More
SHP Steriltechnik AG SegoSoft™ Software
Artikelnummer. 31908789
Ansicht ändern
Click to view available options
Zur Verwendung mit (Geräte):
Autoclaves
Packungsgröße:
1 Stück
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Artikelnummer. Zur Verwendung mit (Geräte) Packungsgröße
31908789 Autoclaves 1 Stück
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Artikelnummer. 31908789 Lieferant SHP Steriltechnik AG Lieferanten-Nr. 30.0000.600.000
 Eingeschränkter Artikel
Verfügbar ab: 12-11-2026
Zum Warenkorb hinzufügen
Zum Warenkorb hinzufügen
missing translation for 'validMainframeMsg'
SegoSoft™ is a Windows‐based software solution designed for fully automatic digital documentation and traceability of instrument reprocessing workflows in medical and laboratory environments.
  • SegoSoft automatically captures and documents processing cycles from connected devices, including start/end times and all relevant cycle data
  • It supports over 200 different devices for sterilisation, cleaning and disinfection, so it works with equipment from a wide range of manufacturers
  • Documentation is stored in a format protected against unauthorized changes and can be archived long‐term for quality management and audit purposes
  • The software includes a unique user administration system based on digital signatures to ensure authenticated approvals and tamper‐proof records
  • It offers barcode label printing for easy traceability of batches and assignment to patient records, often via add‐ons like SegoLabel and SegoAssign
  • SegoSoft integrates into existing practice software systems and workflows, helping reduce manual documentation work and minimise liability risk
Spezifikation
Zur Verwendung mit (Geräte) Autoclaves
Produkttyp Software