Learn More
Secretogranin II Rabbit anti-Human, Unconjugated, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Marke:  Proteintech 20357-1-AP-150UL
 Lernen Sie weitere Varianten dieses
Artikelnummer. 16869884
  • 385.92 CHF / 150 Mikroliter
om dit product te kopen Registreer vandaag om een webaccount aan te maken
Beschreibung
Beschreibung
The Secretogranin II antibody from Proteintech is a rabbit polyclonal antibody generated with fusion protein of human Secretogranin II. This antibody recognizes human, mouse, rat antigen. The Secretogranin II antibody has been validated for the following applications: IF, IHC, WB, ELISA analysis.
Spezifikation
Spezifikation
Secretogranin II
Polyclonal
Unconjugated
150 μL
7857
IgG
Western Blot, Immunohistochemistry, Immunofluorescence, Co-immunoprecipitation (Co-IP), ELISA
233 μg/mL
Rabbit
Primary
Human, Rat, Mouse
Produktvorschläge
Produktvorschläge
Videos
Sicherheitsdatenblatt (SDS)
Dokumentation
Dokumentation
Zertifikate
Aktionen
Aktionen