Learn More
SECISBP2L Polyclonal Antibody, Invitrogen™
Rabbit Polyclonal Antibody
Marke: Thermo Scientific PA558960
Additional Details : Gewicht : 0.02000kg
Beschreibung
Immunogen sequence: EEALKNVKKV PHHMGHSRNP SAASAISFCS VISEPISEVN EKEYETNWRN MVETSDGLEA SENEKEVSCK HSTSEKPSKL PFDTPAIGKQ PSLVATGSTT SATSAGKSTA SDK Highest antigen sequence identity to the following orthologs: Mouse - 73%, Rat - 76%.
SECISBP2L Polyclonal antibody specifically detects SECISBP2L in Human samples. It is validated for Immunocytochemistry, Immunohistochemistry (Paraffin).Spezifikation
SECISBP2L | |
Polyclonal | |
Unconjugated | |
SECISBP2L | |
KIAA0256; SBP2L; SECIS binding protein 2 like; SECIS binding protein 2-like; SECIS-binding protein 2-like; SECISBP2L; Selenocysteine insertion sequence-binding protein 2-like; SLAN | |
Rabbit | |
Antigen affinity chromatography | |
RUO | |
9728 | |
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
Liquid |
Immunocytochemistry, Immunohistochemistry | |
0.2 mg/mL | |
PBS with 40% glycerol and 0.02% sodium azide; pH 7.2 | |
Q93073 | |
SECISBP2L | |
Recombinant protein corresponding to Human SECISBP2L | |
100 μL | |
Primary | |
Human | |
Antibody | |
IgG |
Research Use Only